Assay Details
Target Gene Details
Entrez Gene ID: | 329910 |
Gene Name: | acyl-CoA thioesterase 11 |
Gene Aliases: |
1110020M10Rik, 2010309H15Rik, AW060409, BFIT, BFIT1, Thea, Them1, mKIAA0707 |
Location: |
Chr.4:106744555-106807180 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot11 | NM_025590.4 | 16 | 4184 | NP_079866.2 |
| AK122350.1 | 15 | 3971 | BAC65632.1 | |
| BC042492.1 | 16 | 4119 | AAH42492.1 |
Target Gene Details
Entrez Gene ID: | 230579 |
Gene Name: | family with sequence simliarity 151, member A |
Gene Aliases: |
2010309H15Rik |
Location: |
Chr.4:106733915-106748292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam151a | NM_146149.1 | NP_666261.1 | ||
| AY168619.1 | AAO38748.1 | |||
| BC024426.1 | AAH24426.1 | |||
| BC024430.1 | AAH24430.1 | |||
| BC026682.1 | AAH26682.1 |
More Information
Set Membership: |
|

Genomic Map