Assay Details
Target Gene Details
Entrez Gene ID: | 12804 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
Cntfralpha |
Location: |
Chr.4:41657498-41697091 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntfr | NM_001136056.2 | NP_001129528.1 | ||
| NM_001146080.1 | NP_001139552.1 | |||
| NM_016673.2 | NP_057882.2 | |||
| AB154246.1 | ||||
| AF068615.1 | AAC25711.1 | |||
| AK015675.1 | BAB29926.1 | |||
| AK137026.1 | BAE23207.1 | |||
| BC046974.1 | AAH46974.1 | |||
| BC050928.1 | AAH50928.1 | |||
| BY715214.1 | ||||
| CX202454.1 |
More Information
Set Membership: |
|

Genomic Map