Assay Details
Target Gene Details
Entrez Gene ID: | 72479 |
Gene Name: | hydroxysteroid dehydrogenase like 2 |
Gene Aliases: |
2610207I16Rik |
Location: |
Chr.4:59580458-59618694 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsdl2 | NM_001301367.1 | NP_001288296.1 | ||
| NM_024255.3 | NP_077217.2 | |||
| AJ293845.1 | CAC34587.1 | |||
| AK011910.1 | BAB27910.1 | |||
| AK085899.1 | BAC39563.1 | |||
| AK136773.1 | BAE23126.1 | |||
| AK144948.1 | ||||
| AK145230.1 | BAE26313.1 | |||
| AY725196.1 | AAU11505.1 | |||
| BC002211.1 | AAH02211.1 |
More Information
Set Membership: |
|

Genomic Map