Assay Details
Target Gene Details
Entrez Gene ID: | 329872 |
Gene Name: | Fras1 related extracellular matrix protein 1 |
Gene Aliases: |
BC037594, D430009N09, D630008K06, crf11, eye<m02Jus>, eyes2, heb, qbrick |
Location: |
Chr.4:82897920-83052548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frem1 | NM_001198811.1 | NP_001185740.1 | ||
| NM_177863.4 | NP_808531.2 | |||
| AB160986.1 | BAD89014.1 | |||
| AJ616838.1 | CAE83577.1 | |||
| BC088732.1 | AAH88732.1 | |||
| BC138427.1 |

Genomic Map