Assay Details
Target Gene Details
Entrez Gene ID: | 56226 |
Gene Name: | espin |
Gene Aliases: |
je |
Location: |
Chr.4:152120331-152152420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Espn | NM_019585.3 | NP_062531.2 | ||
| NM_207687.2 | NP_997570.1 | |||
| NM_207688.2 | NP_997571.1 | |||
| NM_207689.2 | NP_997572.1 | |||
| NM_207690.2 | NP_997573.1 | |||
| NM_207691.2 | NP_997574.2 | |||
| AF134857.1 | AAD24481.1 | |||
| AF239886.1 | AAF98134.1 | |||
| AF540942.1 | AAO50326.1 | |||
| AF540943.1 | AAO50327.1 | |||
| AF540944.1 | AAO50328.1 | |||
| AF540945.1 | AAO50329.1 | |||
| AK002284.1 | BAB21987.1 | |||
| AK146701.1 | ||||
| AK165010.1 | ||||
| AY587570.1 | AAT46472.1 | |||
| AY587571.1 | AAT46473.1 | |||
| BC044723.1 | AAH44723.2 | |||
| BC057888.1 | AAH57888.1 | |||
| CJ194793.1 |
More Information
Set Membership: |
|

Genomic Map