Assay Details
Target Gene Details
Entrez Gene ID: | 57741 |
Gene Name: | nucleolar complex associated 2 homolog (S. cerevisiae) |
Gene Aliases: |
AA410003, AF155546, NIR |
Location: |
Chr.4:156235823-156247616 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Noc2l |
