Assay Details
Target Gene Details
Entrez Gene ID: | 14916 |
Gene Name: | guanylate cyclase activator 2b (retina) |
Gene Aliases: |
AV066530, Gcap2, Ugn |
Location: |
Chr.4:119656601-119658945 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Guca2b | NM_008191.2 | 3 | 475 | NP_032217.2 |
| AK002364.1 | 3 | 447 | BAB22042.1 | |
| AV002130.1 | 2 | 228 | ||
| AV003693.1 | 1 | 107 | ||
| BC024373.1 | 3 | 442 | AAH24373.1 | |
| CF423670.1 | 3 | 488 | ||
| U67800.1 | 3 | 431 | AAD09215.1 | |
| U90727.1 | 3 | 477 | AAB53314.1 |
More Information
Set Membership: |
|

Genomic Map