Assay Details
Target Gene Details
Entrez Gene ID: | 71957 |
Gene Name: | cleavage and polyadenylation specific factor 3-like |
Gene Aliases: |
2410006F12Rik, AU044035 |
Location: |
Chr.4:155869548-155889103 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf3l | NM_028020.3 | NP_082296.1 | ||
| AK010425.1 | BAB26928.1 | |||
| AK090206.1 | BAC41135.1 | |||
| AK150436.1 | BAE29558.1 | |||
| AK152740.1 | BAE31460.1 | |||
| AK167607.1 | BAE39661.1 | |||
| AK172533.1 | BAE43054.1 | |||
| BC011155.1 | AAH11155.1 |
More Information
Set Membership: |
|

Genomic Map