Assay Details
Target Gene Details
Entrez Gene ID: | 13200 |
Gene Name: | dolichyl-di-phosphooligosaccharide-protein glycotransferase |
Gene Aliases: |
- |
Location: |
Chr.4:138304738-138312611 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddost | NM_007838.2 | NP_031864.2 | ||
| AK077651.1 | ||||
| AK081542.1 | BAC38250.1 | |||
| AK084267.1 | ||||
| AK148095.1 | BAE28341.1 | |||
| AK150683.1 | BAE29763.1 | |||
| AK196638.1 | ||||
| AK200087.1 | ||||
| BC068132.1 | AAH68132.1 | |||
| D89063.1 | BAA23671.1 |
More Information
Set Membership: |
|

Genomic Map