Assay Details
Target Gene Details
Entrez Gene ID: | 13200 |
Gene Name: | dolichyl-di-phosphooligosaccharide-protein glycotransferase |
Gene Aliases: |
- |
Location: |
Chr.4:138304738-138312611 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddost | NM_007838.2 | 9 | 1141 | NP_031864.2 |
| AK077651.1 | 9 | 1003 | ||
| AK081542.1 | 9 | 1033 | BAC38250.1 | |
| AK084267.1 | 9 | 1025 | ||
| AK148095.1 | 9 | 1022 | BAE28341.1 | |
| AK150683.1 | 9 | 1032 | BAE29763.1 | |
| AK177159.1 | 2 | 103 | ||
| BC068132.1 | 9 | 1141 | AAH68132.1 | |
| D89063.1 | 9 | 1143 | BAA23671.1 |
More Information
Set Membership: |
|

Genomic Map