Assay Details
Target Gene Details
Entrez Gene ID: | 54709 |
Gene Name: | eukaryotic translation initiation factor 3, subunit I |
Gene Aliases: |
36kDa, D4Ertd632e, Eif3s2, Trip1 |
Location: |
Chr.4:129591974-129600648 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3i | NM_018799.2 | NP_061269.1 | ||
| AK149955.1 | ||||
| AK172208.1 | BAE42882.1 | |||
| CJ079467.1 |
Target Gene Details
Entrez Gene ID: | 76799 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
1500002D11Rik, 2510006D16Rik, 4933407D05Rik |
Location: |
Chr.4:129600691-129608528 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem234 | NM_029748.2 | NP_084024.1 | ||
| AK005111.1 | ||||
| AK016715.1 | BAB30394.1 | |||
| AK050648.1 | BAC34362.1 | |||
| AK171961.1 | ||||
| BC024696.1 | AAH24696.1 |
More Information
Set Membership: |
|

Genomic Map