Assay Details
Target Gene Details
Entrez Gene ID: | 68180 |
Gene Name: | hydroxypyruvate isomerase homolog (E. coli) |
Gene Aliases: |
2700033B16Rik, 6430559E15Rik, HT036 |
Location: |
Chr.4:118359999-118362744 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hyi | NM_026601.3 | 1 | 207 | NP_080877.3 |
| NR_003946.2 | 1 | 207 | ||
| AA403499.1 | 1 | 219 | ||
| AK020099.1 | 1 | 193 | ||
| AK090361.1 | 1 | 190 | ||
| BC024619.1 | 1 | 195 | AAH24619.1 | |
| BC100401.1 | 1 | 171 | AAI00402.1 | |
| CK620463.1 | 1 | 207 |
More Information
Set Membership: |
|

Genomic Map