Assay Details
Target Gene Details
Entrez Gene ID: | 68180 |
Gene Name: | hydroxypyruvate isomerase homolog (E. coli) |
Gene Aliases: |
2700033B16Rik, 6430559E15Rik, HT036 |
Location: |
Chr.4:118359999-118362744 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hyi | NM_026601.3 | NP_080877.3 | ||
| NR_003946.2 | ||||
| AK004328.1 | ||||
| AK020099.1 | ||||
| BC024619.1 | AAH24619.1 | |||
| BC100401.1 | AAI00402.1 | |||
| BX525112.1 | ||||
| CB952524.1 |
More Information
Set Membership: |
|

Genomic Map