Assay Details
Target Gene Details
Entrez Gene ID: | 57295 |
Gene Name: | isoprenylcysteine carboxyl methyltransferase |
Gene Aliases: |
1700008E11Rik, C80758, Gm13095, HSTE14, OTTMUSG00000010406, PPMT, STE14, pcCMT |
Location: |
Chr.4:152297214-152307126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Icmt | NM_133788.2 | 1 | 244 | NP_598549.1 |
| NR_037908.1 | 1 | 244 | ||
| AF209926.1 | AAG48552.1 | |||
| AK154135.1 | 1 | 176 | BAE32399.1 | |
| AK154410.1 | 1 | 158 | ||
| BC057132.1 | 1 | 154 | AAH57132.1 | |
| BC110695.1 | 1 | 231 | AAI10696.1 | |
| BY094017.1 | 1 | 245 |
More Information
Set Membership: |
|

Genomic Map