Assay Details
Target Gene Details
Entrez Gene ID: | 66233 |
Gene Name: | DNA methyltransferase 1-associated protein 1 |
Gene Aliases: |
1500016M21Rik, DNMAP1, DNMTAP1, Dmtap1, mKIAA1425 |
Location: |
Chr.4:117674681-117682321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmap1 | NM_023178.2 | NP_075667.1 | ||
| AB221661.1 | ||||
| AF265229.1 | AAF87080.1 | |||
| AF438610.1 | AAL31640.1 | |||
| AK005270.1 | ||||
| AK012055.1 | BAB27996.3 | |||
| AK136976.1 | 2 | 285 | ||
| BC002321.1 | AAH02321.1 | |||
| BC045160.1 | AAH45160.1 |
More Information
Set Membership: |
|

Genomic Map