Assay Details
Target Gene Details
Entrez Gene ID: | 76804 |
Gene Name: | lysine (K)-specific demethylase 4C |
Gene Aliases: |
2410141F18Rik, AA517467, Gasc1, Jmjd2c |
Location: |
Chr.4:74242497-74405864 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm4c | NM_001172095.1 | NP_001165566.1 | ||
| NM_144787.2 | NP_659036.1 | |||
| AK042792.1 | BAC31369.2 | |||
| AK048092.1 | ||||
| AK053104.1 | BAC35267.1 | |||
| AK081769.1 | BAC38327.1 | |||
| AK144103.1 | BAE25700.1 | |||
| AK144890.1 | BAE26118.1 | |||
| AK173023.1 | BAD32301.1 | |||
| BC020180.1 | AAH20180.1 | |||
| BC042424.1 | AAH42424.1 |
More Information
Set Membership: |
|

Genomic Map