Assay Details
Target Gene Details
Entrez Gene ID: | 50927 |
Gene Name: | nuclear autoantigenic sperm protein (histone-binding) |
Gene Aliases: |
5033430J04Rik, AI131596, AI317140, D4Ertd767e, Epcs32, Nasp-T |
Location: |
Chr.4:116601052-116627951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nasp | NM_001081475.1 | NP_001074944.1 | ||
| NM_001284229.1 | NP_001271158.1 | |||
| NM_016777.3 | NP_058057.3 | |||
| AF034610.2 | AAB87567.2 | |||
| AF095722.1 | AAC64195.1 | |||
| AK083333.1 | BAC38871.1 | |||
| BC004693.1 | AAH04693.1 |
More Information
Set Membership: |
|

Genomic Map