Assay Details
Target Gene Details
Entrez Gene ID: | 66407 |
Gene Name: | mitochondrial ribosomal protein S15 |
Gene Aliases: |
1500003E24Rik, 2410002B11Rik, Mprs15 |
Location: |
Chr.4:126046928-126055536 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps15 | NM_025544.2 | NP_079820.2 | ||
| AB049947.1 | BAB41000.1 | |||
| AK003126.1 | BAB22586.1 | |||
| AK005131.2 | ||||
| AK008623.1 | BAB25785.1 | |||
| AK010330.1 | BAB26857.1 | |||
| AK050396.1 | BAC34235.1 | |||
| AK140877.1 | ||||
| BC029193.1 | AAH29193.1 | |||
| BC055861.1 | AAH55861.1 | |||
| CF582282.1 |
More Information
Set Membership: |
|

Genomic Map