Assay Details
Target Gene Details
Entrez Gene ID: | 230721 |
Gene Name: | poly(A) binding protein, cytoplasmic 4 |
Gene Aliases: |
- |
Location: |
Chr.4:123262387-123298832 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pabpc4 | NM_130881.2 | NP_570951.2 | ||
| NM_148917.2 | NP_683717.1 | |||
| BC003283.1 | AAH03283.1 | |||
| BC010345.1 | AAH10345.1 | |||
| BC056432.1 | AAH56432.1 |

Genomic Map