Assay Details
Target Gene Details
Entrez Gene ID: | 230233 |
Gene Name: | inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein |
Gene Aliases: |
3110040G09Rik, 6030413P05, C78473, Elp1, IKAP |
Location: |
Chr.4:56749680-56802331 on Build GRCm38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkap | NM_026079.3 | 35 | 3965 | NP_080355.2 |
| AF140786.1 | 35 | 3893 | AAO15309.1 | |
| AF367244.1 | 35 | 3918 | AAL36025.1 | |
| AF387811.1 | 35 | 3932 | AAL40925.1 | |
| AK004402.1 | 3 | 310 | BAB23291.1 | |
| AK031367.1 | 35 | 3910 | ||
| AK032563.1 | 2 | 134 | ||
| AK076152.1 | 8 | 667 | BAC36221.1 | |
| AK147153.1 | 35 | 3968 | BAE27720.1 | |
| AK165168.1 | 32 | 3442 | ||
| AK170341.1 | 1 | 91 | BAE41731.1 | |
| BC052387.1 | 35 | 3877 | AAH52387.2 | |
| BC054468.1 | 6 | 881 | AAH54468.1 |
More Information
Set Membership: |
|

Genomic Map