Assay Details
Target Gene Details
Entrez Gene ID: | 18198 |
Gene Name: | muscle, skeletal, receptor tyrosine kinase |
Gene Aliases: |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
Location: |
Chr.4:58285640-58377848 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Musk | NM_001037127.2 | NP_001032204.2 | ||
| NM_001037128.1 | NP_001032205.1 | |||
| NM_001037129.1 | NP_001032206.1 | |||
| NM_001037130.1 | NP_001032207.1 | |||
| NM_001165996.1 | NP_001159468.1 | |||
| NM_010944.2 | NP_035074.2 | |||
| AB691772.1 | ||||
| AY360453.1 | AAR12275.1 | |||
| AY955296.1 | AAY40318.1 | |||
| AY955297.1 | AAY40319.1 | |||
| AY955298.1 | AAY40320.1 | |||
| AY956403.1 | AAY40321.1 | |||
| AY956404.1 | AAY40322.1 | |||
| BC100342.1 | AAI00343.1 | |||
| U37708.1 | AAA79203.1 | |||
| U37709.1 | AAA79204.1 | |||
| X86444.1 | CAA60165.1 |
More Information
Set Membership: |
|

Genomic Map