Assay Details
Target Gene Details
Entrez Gene ID: | 68493 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4 |
Gene Aliases: |
1110007M04Rik, 3000003G13Rik, AW214064 |
Location: |
Chr.4:24898083-24905001 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf4 | NM_026742.4 | NP_081018.1 | ||
| AK003553.2 | BAB22852.1 | |||
| AK034728.1 | BAC28809.1 | |||
| AK160341.1 | BAE35747.1 | |||
| BC028764.1 | AAH28764.1 | |||
| BY163651.1 |

Genomic Map