Assay Details
Target Gene Details
Entrez Gene ID: | 230103 |
Gene Name: | natriuretic peptide receptor 2 |
Gene Aliases: |
GC-B, GC-B2, GC-B3, cn, mNPR-B, pwe |
Location: |
Chr.4:43630332-43651437 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npr2 | NM_173788.3 | 13 | 2182 | NP_776149.1 |
| AK158195.1 | 8 | 733 | BAE34402.1 | |
| BC042470.1 | 13 | 2182 | AAH42470.1 |
More Information
Set Membership: |
|

Genomic Map