Assay Details
Target Gene Details
Entrez Gene ID: | 230767 |
Gene Name: | IQ motif containing C |
Gene Aliases: |
BC026978 |
Location: |
Chr.4:129615617-129619213 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcc | NM_198026.2 | NP_932143.2 | ||
| AK083217.1 | 1 | 586 | ||
| BY123003.1 |

Genomic Map