Assay Details
Target Gene Details
Entrez Gene ID: | 18198 |
Gene Name: | muscle, skeletal, receptor tyrosine kinase |
Gene Aliases: |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
Location: |
Chr.4:58285640-58377848 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Musk | NM_001037127.2 | 15 | 2038 | NP_001032204.2 |
| NM_001037128.1 | 15 | 1993 | NP_001032205.1 | |
| NM_001037129.1 | 14 | 1969 | NP_001032206.1 | |
| NM_001037130.1 | 13 | 1939 | NP_001032207.1 | |
| NM_010944.2 | 14 | 1963 | NP_035074.2 | |
| AB691772.1 | 13 | 1789 | ||
| AY955296.1 | 15 | 1888 | AAY40318.1 | |
| AY955297.1 | 15 | 1843 | AAY40319.1 | |
| AY955298.1 | 14 | 1819 | AAY40320.1 | |
| AY956403.1 | 14 | 1813 | AAY40321.1 | |
| AY956404.1 | 13 | 1789 | AAY40322.1 | |
| BC100342.1 | 14 | 1893 | AAI00343.1 | |
| U37708.1 | 13 | 1937 | AAA79203.1 | |
| U37709.1 | 14 | 1961 | AAA79204.1 | |
| X86444.1 | 13 | 1867 | CAA60165.1 |
More Information
Set Membership: |
|

Genomic Map