Assay Details
Target Gene Details
Entrez Gene ID: | 100038882 |
Gene Name: | ISG15 ubiquitin-like modifier |
Gene Aliases: |
G1p2, IGI15, IP17, Irfp, UCRP |
Location: |
Chr.4:156199424-156200818 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Isg15 | NM_015783.3 | NP_056598.2 | ||
| BC031424.1 | AAH31424.1 | |||
| BC083156.1 | AAH83156.1 | |||
| BC109346.2 | AAI09347.1 | |||
| X56602.1 | CAA39939.1 |

Genomic Map