Assay Details
Target Gene Details
Entrez Gene ID: | 59001 |
Gene Name: | polymerase (DNA directed), epsilon 3 (p17 subunit) |
Gene Aliases: |
1810034K18Rik, YBL1 |
Location: |
Chr.4:62523798-62525014 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pole3 | NM_021498.2 | NP_067473.2 | ||
| AF230806.1 | AAF67146.1 | |||
| BC024996.1 | AAH24996.1 |
More Information
Set Membership: |
|

Genomic Map