Assay Details
Target Gene Details
Entrez Gene ID: | 64424 |
Gene Name: | polymerase (RNA) I polypeptide E |
Gene Aliases: |
53kDa, AU042259, D030019D19Rik, Paf53, Praf1 |
Location: |
Chr.4:45018609-45034279 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr1e | NM_001285800.1 | 13 | 1287 | NP_001272729.1 |
| NM_022811.3 | 11 | 1143 | NP_073722.1 | |
| AK083948.1 | 11 | 1113 | ||
| AK132556.1 | 11 | 1114 | BAE21232.1 | |
| BC034783.1 | 13 | 1234 | AAH34783.1 | |
| D14336.1 | 11 | 1106 | BAA03266.1 |
More Information
Set Membership: |
|

Genomic Map