Assay Details
Target Gene Details
Entrez Gene ID: | 70603 |
Gene Name: | mutY homolog (E. coli) |
Gene Aliases: |
5730495A01Rik, Mutyha, Mutyhalpha, Mutyhb, Mutyhbeta, Mutyhc, Myh |
Location: |
Chr.4:116807682-116819439 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mutyh | NM_001159581.1 | NP_001153053.1 | ||
| NM_133250.2 | NP_573513.2 | |||
| AB117937.1 | BAC98379.1 | |||
| AB117938.1 | BAC98380.1 | |||
| AB117939.1 | BAC98381.1 | |||
| AK017730.1 | ||||
| AK077546.1 | ||||
| AK156790.1 | BAE33857.1 | |||
| AY007717.1 | AAG16632.1 | |||
| BC057942.1 | AAH57942.1 | |||
| CX213088.1 |
More Information
Set Membership: |
|

Genomic Map