Assay Details
Target Gene Details
Entrez Gene ID: | 18405 |
Gene Name: | orosomucoid 1 |
Gene Aliases: |
Agp-1, Agp-2, Orm-1 |
Location: |
Chr.4:63344566-63348162 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orm1 | NM_008768.1 | NP_032794.1 | ||
| AK138985.1 | ||||
| BC012725.1 | AAH12725.1 | |||
| CT010399.1 | CAJ18605.1 | |||
| M12567.1 | AAA91745.1 | |||
| M27008.1 | AAA37194.1 |

Genomic Map