Assay Details
Target Gene Details
Entrez Gene ID: | 72656 |
Gene Name: | integrator complex subunit 8 |
Gene Aliases: |
2810013E07Rik, AV063769, D130008D20Rik |
Location: |
Chr.4:11199158-11254266 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ints8 | NM_001159595.1 | 17 | 2273 | NP_001153067.1 |
| NM_178112.5 | 17 | 2273 | NP_835213.3 | |
| AK047035.1 | 1 | 864 | ||
| AK049513.1 | 17 | 2244 | BAC33786.1 | |
| AK051161.1 | 1 | 1719 | ||
| AK140829.1 | 17 | 2268 | BAE24491.1 | |
| AK169552.1 | 17 | 2242 | BAE41225.1 | |
| AK176935.1 | ||||
| BC049092.1 | 17 | 2203 | AAH49092.1 | |
| BC050192.1 | 8 | 987 | AAH50192.1 | |
| BC057618.1 | 17 | 2203 | AAH57618.1 |

Genomic Map