Assay Details
Target Gene Details
Entrez Gene ID: | 21687 |
Gene Name: | endothelial-specific receptor tyrosine kinase |
Gene Aliases: |
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2 |
Location: |
Chr.4:94739289-94874976 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tek | NM_001290549.1 | 3 | 758 | NP_001277478.1 |
| NM_001290551.1 | 3 | 758 | NP_001277480.1 | |
| NM_013690.3 | 3 | 758 | NP_038718.2 | |
| BC050824.1 | 3 | 752 | AAH50824.1 | |
| D13738.1 | 3 | 757 | BAA02883.1 | |
| S67051.1 | 4 | 421 | AAB28663.1 | |
| X67553.1 | 3 | 529 | CAA47857.1 | |
| X71426.1 | 4 | 492 | CAA50557.1 |
More Information
Set Membership: |
|

Genomic Map