Assay Details
Target Gene Details
Entrez Gene ID: | 68276 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
4930584N22Rik, 4933424D16Rik, AI413517 |
Location: |
Chr.4:116804003-116807635 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Toe1 | NM_026654.2 | NP_080930.1 | ||
| AK019829.1 | BAB31871.1 | |||
| AK046576.1 | BAC32792.1 | |||
| AK082975.1 | BAC38718.1 | |||
| BC023109.1 | AAH23109.1 |
More Information
Set Membership: |
|

Genomic Map