Assay Details
Target Gene Details
Entrez Gene ID: | 106021 |
Gene Name: | topoisomerase I binding, arginine/serine-rich |
Gene Aliases: |
AW105885, LUN, TP53BPL, p53BP3/LUN |
Location: |
Chr.4:40259606-40269841 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Topors | NM_134097.3 | 1 | 40 | NP_598858.2 |
| AB072395.1 | BAB69457.1 | |||
| AB104865.1 | BAC65157.1 | |||
| AK044564.1 | BAC31981.2 | |||
| AK134075.1 | BAE22003.1 | |||
| AK140250.1 | 1 | 41 | BAE24298.1 | |
| AK143025.1 | BAE25253.1 | |||
| AK153743.1 | BAE32164.1 | |||
| BC019421.1 | ||||
| BC037141.1 | AAH37141.1 | |||
| BC040797.1 | AAH40797.1 |
Target Gene Details
Entrez Gene ID: | 100504309 |
Gene Name: | topoisomerase I binding, arginine/serine-rich, opposite strand |
Gene Aliases: |
2010003O02Rik |
Location: |
Chr.4:40269579-40270221 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Toporsos | NR_045265.1 | |||
| AA614943.1 | ||||
| AK019894.1 |
More Information
Set Membership: |
|

Genomic Map