Assay Details
Target Gene Details
Entrez Gene ID: | 100340 |
Gene Name: | sphingomyelin phosphodiesterase, acid-like 3B |
Gene Aliases: |
1110054A24Rik, AU045240, Asml3b |
Location: |
Chr.4:132732966-132757171 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smpdl3b | NM_133888.2 | 7 | 1065 | NP_598649.1 |
| AK136602.1 | 6 | 727 | ||
| AK154599.1 | 7 | 1091 | BAE32702.1 | |
| AK166253.1 | 7 | 1041 | BAE38663.1 | |
| BC009087.1 | 7 | 1065 | AAH09087.1 |
More Information
Set Membership: |
|

Genomic Map