Assay Details
Target Gene Details
Entrez Gene ID: | 13706 |
Gene Name: | chymotrypsin-like elastase family, member 2A |
Gene Aliases: |
Ela-2, Ela2, Ela2a |
Location: |
Chr.4:141814963-141826003 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cela2a | NM_007919.2 | NP_031945.1 | ||
| AK131813.1 | ||||
| AK160448.1 | ||||
| BC026552.1 | AAH26552.1 | |||
| X04573.1 | CAA28242.1 |
More Information
Set Membership: |
|

Genomic Map