Assay Details
Target Gene Details
Entrez Gene ID: | 13706 |
Gene Name: | chymotrypsin-like elastase family, member 2A |
Gene Aliases: |
Ela-2, Ela2, Ela2a |
Location: |
Chr.4:141814963-141826003 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cela2a | NM_007919.2 | 5 | 447 | NP_031945.1 |
| BC026552.1 | 5 | 436 | AAH26552.1 | |
| X04573.1 | 5 | 447 | CAA28242.1 |
More Information
Set Membership: |
|

Genomic Map