Assay Details
Target Gene Details
Entrez Gene ID: | 12371 |
Gene Name: | caspase 9 |
Gene Aliases: |
AI115399, APAF-3, AW493809, CASP-9, Caspase-9, ICE-LAP6, Mch6 |
Location: |
Chr.4:141793612-141815978 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp9 | NM_001277932.1 | 8 | 2907 | NP_001264861.1 |
| NM_015733.5 | 9 | 3017 | NP_056548.2 | |
| AK085152.1 | 9 | 2773 | BAC39378.1 | |
| AK154495.1 | 9 | 2787 | ||
| BC056372.1 | 9 | 2799 | AAH56372.1 | |
| BC056447.1 | 9 | 3017 | AAH56447.1 |
Target Gene Details
Entrez Gene ID: | 13706 |
Gene Name: | chymotrypsin-like elastase family, member 2A |
Gene Aliases: |
Ela-2, Ela2, Ela2a |
Location: |
Chr.4:141814963-141826003 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cela2a | NM_007919.2 | NP_031945.1 | ||
| BC026552.1 | AAH26552.1 | |||
| X04573.1 | CAA28242.1 |
More Information
Set Membership: |
|

Genomic Map