Assay Details
Target Gene Details
Entrez Gene ID: | 19934 |
Gene Name: | ribosomal protein L22 |
Gene Aliases: |
2700038K18Rik |
Location: |
Chr.4:152324436-152334082 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl22 | NM_001277113.1 | NP_001264042.1 | ||
| NM_001277114.1 | NP_001264043.1 | |||
| NM_009079.3 | NP_033105.1 | |||
| NR_102291.1 | ||||
| NR_102292.1 | ||||
| AA755840.1 | ||||
| AK003022.1 | ||||
| AK009094.1 | ||||
| AK168353.1 | BAE40290.1 | |||
| AK169078.1 | BAE40863.1 | |||
| AK169731.1 | ||||
| BC007139.1 | AAH07139.1 | |||
| BC021344.1 | AAH21344.1 | |||
| BC096390.1 | AAH96390.1 | |||
| D17653.1 | BAA04546.1 |
More Information
Set Membership: |
|

Genomic Map