Assay Details
Target Gene Details
Entrez Gene ID: | 208890 |
Gene Name: | solute carrier family 26, member 7 |
Gene Aliases: |
- |
Location: |
Chr.4:14502430-14621971 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a7 | NM_145947.2 | 3 | 382 | NP_666059.2 |
| AF345194.1 | 2 | 284 | AAO49172.1 | |
| AK054327.1 | 3 | 383 | ||
| BC026928.1 | 3 | 361 | AAH26928.1 |
More Information
Set Membership: |
|

Genomic Map