Assay Details
Target Gene Details
Entrez Gene ID: | 107568 |
Gene Name: | WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
8030445B08Rik, AIP5, SDRP1, Tiul1 |
Location: |
Chr.4:19608296-19709004 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wwp1 | NM_001276292.1 | NP_001263221.1 | ||
| NM_177327.5 | NP_796301.2 | |||
| NR_074089.1 | ||||
| NR_074090.1 | ||||
| AK031762.1 | ||||
| AK032232.1 | ||||
| AK033138.1 | BAC28168.1 | |||
| AK137588.1 | BAE23421.1 | |||
| AK168567.1 | ||||
| BC021470.1 | AAH21470.1 | |||
| BC026829.1 | ||||
| BC055937.1 | AAH55937.1 | |||
| BC150812.1 |
More Information
Set Membership: |
|

Genomic Map