Assay Details
Target Gene Details
Entrez Gene ID: | 433700 |
Gene Name: | sperm associated antigen 8 |
Gene Aliases: |
MH-SPAG8 |
Location: |
Chr.4:43651330-43654124 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag8 | NM_001007463.1 | NP_001007464.1 | ||
| NM_001290462.1 | NP_001277391.1 | |||
| AK132968.1 | BAE21448.1 | |||
| AY769082.1 | AAV31155.1 | |||
| BC150868.1 |
More Information
Set Membership: |
|

Genomic Map