Assay Details
Target Gene Details
Entrez Gene ID: | 67615 |
Gene Name: | ubiquitin-conjugating enzyme E2R 2 |
Gene Aliases: |
1200003M11Rik, Cdc34b, Ubc3b |
Location: |
Chr.4:41136021-41193370 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2r2 | NM_026275.4 | 1 | 153 | NP_080551.1 |
| AJ240086.1 | 1 | 112 | CAC80335.1 | |
| AK003550.1 | 1 | 174 | BAB22850.1 | |
| AK007517.1 | 1 | 286 | BAB25085.1 | |
| AK075703.1 | 1 | 163 | BAC35899.1 | |
| AK075714.1 | 1 | 154 | BAC35904.1 | |
| BC011112.1 | 1 | 158 | AAH11112.1 |

Genomic Map