Assay Details
Target Gene Details
Entrez Gene ID: | 22608 |
Gene Name: | Y box protein 1 |
Gene Aliases: |
1700102N10Rik, C79409, EF1A, MSY1, Nsep1, YB-1, dbpB, mYB-1a |
Location: |
Chr.4:119277327-119294659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx1 | NM_011732.2 | NP_035862.2 | ||
| AF038994.1 | AAB94768.1 | |||
| AK150821.1 | BAE29883.1 | |||
| AK187199.1 | ||||
| AK188049.1 | ||||
| AK205911.1 | ||||
| AK209866.1 | ||||
| AK212436.1 | ||||
| BC013450.1 | AAH13450.1 | |||
| BC013620.1 | AAH13620.1 | |||
| BC029747.1 | AAH29747.1 | |||
| BC031472.1 | AAH31472.1 | |||
| BC049977.1 | AAH49977.1 | |||
| BC061634.1 | AAH61634.1 | |||
| BC106143.1 | AAI06144.1 | |||
| BU512458.1 | ||||
| M60419.1 | AAA40577.1 | |||
| M62867.1 | AAA63390.1 | |||
| U33196.1 | AAA75475.1 | |||
| U33197.1 | AAA75476.1 | |||
| X57621.1 | CAA40847.1 |

Genomic Map