Assay Details
Target Gene Details
Entrez Gene ID: | 54170 |
Gene Name: | Ras-related GTP binding C |
Gene Aliases: |
AU041672, Gtr2, RAGC, TIB929, YGR163W |
Location: |
Chr.4:123917433-123936997 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rragc | NM_017475.2 | NP_059503.2 | ||
| AB017616.1 | BAA75671.1 | |||
| AK150792.1 | ||||
| AK159355.1 | BAE35013.1 | |||
| AK159434.1 | ||||
| AK166658.1 | BAE38923.1 | |||
| BC005417.1 | AAH05417.1 | |||
| BC071245.1 | 2 | 487 | AAH71245.1 |
More Information
Set Membership: |
|

Genomic Map