Assay Details
Target Gene Details
Entrez Gene ID: | 73723 |
Gene Name: | SH3 domain binding glutamic acid-rich protein-like 3 |
Gene Aliases: |
1110004L05Rik, AU022188 |
Location: |
Chr.4:134127406-134128753 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3bgrl3 | NM_080559.1 | NP_542126.1 | ||
| AK003431.1 | BAC25034.1 | |||
| AK179341.1 | ||||
| BC008110.1 | AAH08110.1 |

Genomic Map