Assay Details
Target Gene Details
Entrez Gene ID: | 58212 |
Gene Name: | serine/arginine repetitive matrix 3 |
Gene Aliases: |
2900083I11Rik, AB041661, SRm300-like, Srrm2l |
Location: |
Chr.5:135806861-135874773 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrm3 | NM_021403.3 | NP_067378.2 | ||
| BC051947.1 | AAH51947.1 |

Genomic Map