Assay Details
Target Gene Details
Entrez Gene ID: | 230979 |
Gene Name: | tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator) |
Gene Aliases: |
Atar, HveA, Hvem, Tnfrs14 |
Location: |
Chr.4:154921442-154928620 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf14 | NM_178931.2 | NP_849262.1 | ||
| AF515707.1 | AAQ08183.1 | |||
| AY264405.1 | AAO89081.1 | |||
| BC022125.1 | AAH22125.1 | |||
| BC104054.1 | AAI04055.1 | |||
| BC104055.1 | AAI04056.1 |
More Information
Set Membership: |
|

Genomic Map