Assay Details
Target Gene Details
Entrez Gene ID: | 70920 |
Gene Name: | RIKEN cDNA 4921511H03 gene |
Gene Aliases: |
- |
Location: |
Chr.5:7302505-7311491 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4921511H03Rik | AK014870.1 | BAB29593.1 | ||
| AK133311.1 | BAE21604.1 | |||
| BC049718.1 | AAH49718.1 |
Target Gene Details
Entrez Gene ID: | 207618 |
Gene Name: | zinc finger protein 804B |
Gene Aliases: |
Gm441 |
Location: |
Chr.5:6769030-7344378 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp804b | NM_001163223.1 | NP_001156695.1 |
More Information
Set Membership: |
|

Genomic Map