Assay Details
Target Gene Details
Entrez Gene ID: | 242773 |
Gene Name: | solute carrier family 45, member 1 |
Gene Aliases: |
C230078B22, Dnb5 |
Location: |
Chr.4:150629396-150652174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc45a1 | NM_173774.3 | NP_776135.2 | ||
| AK134677.1 | 8 | 2067 | ||
| AK134683.1 |
More Information
Set Membership: |
|

Genomic Map